missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serum Amyloid A (SAA1) Monoclonal Antibody (OTI7A9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies CF814058
This item is not returnable.
View return policy
Description
Serum Amyloid A (SAA1) Monoclonal antibody specifically detects Serum Amyloid A (SAA1) in Human samples. It is validated for ELISASpecifications
| Serum Amyloid A (SAA1) | |
| Monoclonal | |
| See Label | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P0DJI8 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 6288 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| ELISA | |
| OTI7A9 | |
| Unconjugated | |
| SAA1 | |
| amyloid a; Amyloid fibril protein AA; Amyloid protein A; MGC111216; PIG4; SAA; SAA1; Saa-1; Saa2; serum amyloid A; serum amyloid A 1; serum amyloid A 2; serum amyloid A protein; serum amyloid A1; Serum amyloid A1 protein; Serum amyloid A-1 protein; serum amyloid A-like protein; Serum amyloid protein A(2-102); Serum amyloid protein A(2-103); Serum amyloid protein A(2-104); Serum amyloid protein A(3-104); Serum amyloid protein A(4-101); Serum Amyloid-A; TP53I4; tumor protein p53 inducible protein 4 | |
| Human recombinant protein fragment corresponding to amino acids 19-122 of human SAA (NP_954630) produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction