missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serum Amyloid A1/A2 Antibody (SAA/2868R), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08413H
This item is not returnable.
View return policy
Description
Serum Amyloid A1/A2 Monoclonal antibody specifically detects Serum Amyloid A1/A2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Serum Amyloid A1/A2 | |
| Monoclonal | |
| HRP | |
| PIG4;SAA;SAA1;SAA2;Serum amyloid A-1 protein;Serum amyloid A-2 protein;TP53I4 | |
| Recombinant full-length human Serum Amyloid A1/A2 (SAA) protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| SAA/2868R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 6288 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction