missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serum Amyloid A1 Antibody (SAA1cc), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-73071AF647
This item is not returnable.
View return policy
Description
Serum Amyloid A1 Monoclonal antibody specifically detects Serum Amyloid A1 in Human, Canine samples. It is validated for Western Blot, ELISA
Specifications
| Serum Amyloid A1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| MGC111216, PIG4, SAA, SAA2, serum amyloid A protein, serum amyloid A1, TP53I4, tumor protein p53 inducible protein 4 | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Human SAA. | |
| 0.2 mL | |
| Alzheimers Research, Cancer, Neuroscience | |
| 6288 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, ELISA | |
| SAA1cc | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction