missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serum Amyloid A4 Antibody (126), Biotin , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89888B
This item is not returnable.
View return policy
Description
Serum Amyloid A4 Monoclonal antibody specifically detects Serum Amyloid A4 in Human samples. It is validated for ELISA
Specifications
| Serum Amyloid A4 | |
| Monoclonal | |
| Biotin | |
| C-SAACSAAConstitutively expressed serum amyloid A protein, serum amyloid A-4 protein, serum amyloid A4, constitutive | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Serum Amyloid A4 (P35542; Glu19-Tyr130). | |
| 0.1 mL | |
| Immunology | |
| 6291 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 126 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto