missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serum Response Factor SRF Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71927F
This item is not returnable.
View return policy
Description
Serum Response Factor SRF Polyclonal antibody specifically detects Serum Response Factor SRF in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Serum Response Factor SRF | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| MCM1, serum response factor, serum response factor (c-fos serum response element-binding transcriptionfactor) | |
| The immunogen for this product maps to a region between residue 25 and 75 of human Serum Response Factor using the numbering given in entry NP_003122.1 (GeneID 6722). | |
| 0.1 mL | |
| Phospho Specific | |
| 6722 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| FITC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction