missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SESN1 Rabbit anti-Human, Mouse, Rat, Clone: 7I5Y4, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 207.00 - € 497.00
Specifications
| Antigen | SESN1 |
|---|---|
| Clone | 7I5Y4 |
| Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18054835
|
Novus Biologicals
NBP3-15916-20UL |
20 μg |
€ 207.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18358174
|
Novus Biologicals
NBP3-15916-100UL |
100 μg |
€ 497.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SESN1 Monoclonal antibody specifically detects SESN1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| SESN1 | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| MGC138241, MGC142129, p53-regulated protein PA26, PA26p53 activated gene 26, SEST1p53 regulated PA26 nuclear protein, sestrin 1, sestrin-1 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human SESN1 (Q9Y6P5). KTQHYLLQMDGPLPLHYRHYIGIMAAARHQCSYLVNLHVNDFLHVGGDPKWLNGLENAPQKLQNLGELNKVLAHRPWLITKEHIEGLLKAEEHSWSLAELV | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 7I5Y4 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 27244 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title