missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SESN2 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-44990
This item is not returnable.
View return policy
Description
SESN2 Polyclonal specifically detects SESN2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications
| SESN2 | |
| Polyclonal | |
| Western Blot >1:500, ELISA < 1:10000 | |
| DKFZp761M0212, DKFZp761M02121, Hi95, HI95sestrin-2, SES2, SEST2hypoxia induced gene 95, sestrin 2 | |
| Rabbit | |
| 54.6 kDa | |
| 0.1 mg | |
| Cell Cycle and Replication | |
| 83667 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA | |
| FITC | |
| Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide | |
| SESN2 | |
| Synthetic peptide taken within amino acid region 1-80 on SESN2 protein. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is specific to SESN2 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction