missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SESTD1 Monoclonal Antibody (OTI3A11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505833
This item is not returnable.
View return policy
Description
SESTD1 was initially identified in mutant zebrafish with defects in the spontaneous contraction and touch response as a novel gene, solo, encoding a protein containing SEC14 and spectrin repeat domains. Other experiments indicated that SESTD1 interacts with the TRPC4 and TRPC5, members of the transient receptor potential channel family, via the TRPC calmodulin- and inositol 1, 4, 5-triphosphate receptor-binding domain and is essential for efficient receptor-mediated activation of TRPC5, suggesting that SESTD1 is a novel regulator of these TRPC proteins.Specifications
| SESTD1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q86VW0 | |
| SESTD1 | |
| Full length human recombit protein of human SESTD1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI3A11 | |
| Unconjugated | |
| SESTD1 | |
| SOLO | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 91404 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction