missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SESTD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585032
This item is not returnable.
View return policy
Description
Immunogen sequence: WNVVKTVVVM LQNVVPAEVS LVCVVKPDEF WDKKVTHFCF WKEKDRLGFE VILVSANKLT RYIEPCQLTE DFGGSL Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 99%.
SESTD1 was initially identified in mutant zebrafish with defects in the spontaneous contraction and touch response as a novel gene, solo, encoding a protein containing SEC14 and spectrin repeat domains. Other experiments indicated that SESTD1 interacts with the TRPC4 and TRPC5, members of the transient receptor potential channel family, via the TRPC calmodulin- and inositol 1, 4, 5-triphosphate receptor-binding domain and is essential for efficient receptor-mediated activation of TRPC5, suggesting that SESTD1 is a novel regulator of these TRPC proteins.
Specifications
| SESTD1 | |
| Polyclonal | |
| Unconjugated | |
| Sestd1 | |
| 1500031J16Rik; A930010O20; EGK_04585; Huntingtin-interacting protein-like protein; protein Solo; RGD1562244; SEC14 and spectrin domain containing 1; SEC14 and spectrin domains 1; SEC14 domain and spectrin repeat-containing protein 1; Sestd1; SOLO; Solo/Trio8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 91404 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86VW0 | |
| Sestd1 | |
| Recombinant protein corresponding to Human SESTD1. Recombinant protein control fragment (Product #RP-108680). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction