Learn More
Abnova™ SET Recombinant Protein
Product Code. 16100682
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. 16100682
Brand: Abnova™ H00006418P01.10ug
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Accession Number AAH32749.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 6418
Molecular Weight (g/mol) 56.21
Name SET (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Source Wheat Germ (in vitro)
Immunogen MSAPAAKVSKKELNSNHDGADETSEKEQQEAIEHIDEVQNEIDRLNEQASEEILKVEQKYNKLRQPFFQKRSELIAKIPNFWVTTFVNHPQVSALLGEEDEEALHYLTRVEVTEFEDIKSGYRIDFYFDENPYFENKVLSKEFHLNESGDPSSKSTEIKWKSGKDLTKRSSQTQNKASRKRQHEEPESFFTWFTDHSDAGADELGEVIKDDIWPNPLQYYLVPDMDDEEGEGEEDDDDDEEEEGLEDIDEEGDEDEGEEDEDDDEGEEGEEDEGEDD
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias 2PP2A/I2PP2A/IGAAD/IPP2A2/PHAPII/TAF-I/TAF-IBETA
Common Name SET
Gene Symbol SET
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution