missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SET1B Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06532H
This item is not returnable.
View return policy
Description
SET1B Polyclonal antibody specifically detects SET1B in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| SET1B | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence | |
| EC 2.1.1, FLJ20803, histone-lysine N-methyltransferase SETD1B, hSET1B, KIAA1076EC 2.1.1.43, KMT2GSET1B, Lysine N-methyltransferase 2G, SET domain containing 1B, SET domain-containing protein 1B, Set1B | |
| Produced in rabbits immunized with E. coli-derived Human SET1B fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry | |
| HRP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 23067 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction