missing translation for 'onlineSavingsMsg'
Learn More

SETD1A Antibody (BLR024E), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP2-76401

Product Code. 18213879

  • € 454.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SETD1A Monoclonal specifically detects SETD1A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SETD1A
Monoclonal
Unconjugated
Borate Buffered Saline (BBS) pH 8.2 with 0.1% BSA with 0.09% Sodium Azide
SETD1A
Immunogen sequence between amino acids 1200 and 1250
100 μL
9739.0
Store at 2-8°C/ 1 year from date of receipt
IgG
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
BLR024E
Western Blot 1:1000, Immunohistochemistry 1:25 - 1:250, Immunocytochemistry/Immunofluorescence 1:25-1:250, Immunoprecipitation 20 ul/mg lysate, Immunohistochemistry-Paraffin
histone-lysine N-methyltransferase SETD1A, hSET1A, KIAA0339EC 2.1.1.43, KMT2FSET1, Lysine N-methyltransferase 2F, SET domain containing 1A, SET domain-containing protein 1A, Set1, Set1/Ash2 histone methyltransferase complex subunit SET1, SET1A
Rabbit
Antigen Affinity-purified
Primary
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.