Learn More
SETD1A Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NB100-558JF646
Product Code. 18710982
  • € 681.00 / 0.10mL
Description
SETD1A Polyclonal antibody specifically detects SETD1A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SETD1A
Polyclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin)
0.1 mL
Store at 4C in the dark.