Learn More
SETD2 Monoclonal Antibody (OTI3G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804268S
This item is not returnable.
View return policy
Description
SETD2 Monoclonal antibody specifically detects SETD2 in Human samples. It is validated for Western BlotSpecifications
| SETD2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q99731 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6363 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI3G8 | |
| Unconjugated | |
| Ccl19 | |
| 4921524K10Rik; BC031601; FLJ16420; FLJ22472; FLJ23184; FLJ45883; HBP231; HIF1; HIF-1; HIP-1; histone-lysine N-methyltransferase SETD2; hSET2; HSPC069; huntingtin interacting protein 1; huntingtin yeast partner B; Huntingtin-interacting protein 1; Huntingtin-interacting protein B; HYPB; Kiaa1732; Kif9; kinesin family member 9; KMT3A; LLS; lysine N-methyltransferase 3A; p231HBP; Protein-lysine N-methyltransferase SETD2; SET domain containing 2; SET domain containing 2, histone lysine methyltransferase; SET domain-containing protein 2; SET2; Setd2 | |
| Human recombinant protein fragment corresponding to amino acids 22-98 of human CCL19 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |