missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SETD5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA557318
This item is not returnable.
View return policy
Description
Immunogen sequence: ETPVGEETKT EAPESEVSNS VSNVTIPSTP QSVGVNTRRS SQAGDIAAEK LVPKPPPAKP SRPRPKSRIS RYRTSSAQRL KRQKQ Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 85%.
Specifications
| SETD5 | |
| Polyclonal | |
| Unconjugated | |
| SETD5 | |
| SETD5 | |
| Recombit protein corresponding to Human SETD5 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9C0A6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55209 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction