Learn More
SETD6 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93031-0.02ml
This item is not returnable.
View return policy
Description
SETD6 Polyclonal antibody specifically detects SETD6 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| SETD6 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| SET domain containing 6, SET domain-containing protein 6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 301-473 of human SETD6 (NP_001153777.1). EPYPDNTDDTADIQMVTVREAALQGTKTEAERHLVYERWDFLCKLEMVGEEGAFVIGREEVLTEEELTTTLKVLCMPAEEFRELKDQDGGGDDKREEGSLTITNIPKLKASWRQLLQNSVLLTLQTYATDLKTDQGLLSNKEVYAKLSWREQQALQVRYGQKMILHQLLELTS | |
| 0.02 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 79918 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |