missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Hepatitis E Virus Antibody (IgG) ELISA Kit (Colorimetric)

Product Code. 18292622 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18292622

Brand: Novus Biologicals™ NBP2600481KIT

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Colorimetric ELISA Kit for study of Immunity, chemokine, Chemotaxis

The Human Hepatitis E Virus Antibody (IgG) ELISA Kit (Colorimetric) measures human Hepatitis E Virus Antibody (IgG) in serum, plasma.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range Qualitative
Product Type ELISA Kit (Colorimetric)
Sample Type Plasma, Serum
Specificity For detection of human hepatitis E virus antibody (IgG)
Target Species Human
For Use With (Application) ELISA
Kit Contents Coated assay plate, Negative Control, Positive Control, Sample Diluents, HRP- conjugate, Wash Buffer (30X concentrate), Substrate A, Substrate B, Stop Solution, Adhesive Strip (For 96 wells), Instruction manual
Quantity 1 Kit
Regulatory Status RUO
Research Discipline Cell Biology
Sample Volume 50 to 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Target Hepatitis E Virus Antibody (IgG)
Show More Show Less

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.