Learn More
SETD7/9 Antibody (7C1B7E1), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-06625MFV610
Product Code. 18566673
  • € 451.00 / 0.10mL
Description
SETD7/9 Monoclonal antibody specifically detects SETD7/9 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SETD7/9
Monoclonal
mFluor Violet 610 SE
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified
ELISA
7C1B7E1
ELISA
EC 2.1.1.43, H3-K4-HMTase SETD7, histone H3-lysine 4-specific methyltransferase, histone-lysine N-methyltransferase SETD7, KIAA1717SET domain-containing protein 7, KMT7FLJ21193, Lysine N-methyltransferase 7, SET domain containing (lysine methyltransferase) 7, SET7/9Histone H3-K4 methyltransferase SETD7, SET7Set9, SET9
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SETD7/9 (Accession#: NP_085151.1; Asp 2-Lys 366).
0.1 mL
Histones and Modified Histones, Signal Transduction
80854
Store at 4C in the dark.
IgG2b