missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SETD7 Antibody (OTI2D10), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74145MFV500
This item is not returnable.
View return policy
Description
SETD7 Monoclonal antibody specifically detects SETD7 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry
Specifications
| SETD7 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| Purified |
| Flow Cytometry | |
| OTI2D10 | |
| Flow Cytometry | |
| EC 2.1.1.43, H3-K4-HMTase SETD7, histone H3-lysine 4-specific methyltransferase, histone-lysine N-methyltransferase SETD7, KIAA1717SET domain-containing protein 7, KMT7FLJ21193, Lysine N-methyltransferase 7, SET domain containing (lysine methyltransferase) 7, SET7/9Histone H3-K4 methyltransferase SETD7, SET7Set9, SET9 | |
| Full length human recombinant protein of human SETD7(NP_085151) produced in HEK293T cell. | |
| 0.1 mL | |
| Epigenetics | |
| 80854 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction