missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SETD8 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-91269AF594
This item is not returnable.
View return policy
Description
SETD8 Polyclonal specifically detects SETD8 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| SETD8 | |
| Polyclonal | |
| Western Blot | |
| H4-K20-HMTase SETD8, H4K20-specific histone methyltransferase splice variant Set8b, Histone-lysine N-methyltransferase SETD8, KMT5A, KMT5A SET domain-containing protein 8, Lysine N-methyltransferase 5A, PR/SET domain containing protein 8, PR/SET domain-containing protein 07, PR/SET07, PRSET7, PR-Set7EC 2.1.1.43, SET domain containing (lysine methyltransferase) 8, SET07SET8H4-K20-specific histone methyltransferase, SET8 | |
| Rabbit | |
| 0.1 mL | |
| 387893 | |
| Store at 4°C in the dark. |
| Western Blot | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| KMT5A | |
| A synthetic peptide made to an internal portion of the human SETD8 protein (between residues 100-200) [UniProt Q9NQR1] | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction