missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SETDB1 Antibody (5H6A12), Novus Biologicals™
Mouse Monoclonal Antibody
€ 230.00
Specifications
| Antigen | SETDB1 |
|---|---|
| Clone | 5H6A12 |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
SETDB1 Monoclonal specifically detects SETDB1 in Human, Mouse, Monkey samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| SETDB1 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Q15047 | |
| 9869 | |
| Purified recombinant fragment of human SETDB1 expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 5H6A12 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| ERG-associated protein with SET domain, ESETEC 2.1.1.43, H3-K9-HMTase 4, Histone H3-K9 methyltransferase 4, histone-lysine N-methyltransferase SETDB1, histone-lysine N-methyltransferase, H3lysine-9 specific 4, KG1T, KIAA0067H3-K9-HMTase4, KMT1EERG-associated protein with a SET domain, ESET, Lysine N-methyltransferase 1E, SET domain bifurcated 1, SET domain, bifurcated 1 | |
| SETDB1 | |
| IgG1 | |
| Unpurified | |
| 180 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title