missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SETDB1 Antibody [PE], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NB100-79776PE
This item is not returnable.
View return policy
Description
SETDB1 Polyclonal antibody specifically detects SETDB1 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| SETDB1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| PE | |
| ERG-associated protein with SET domain, ESETEC 2.1.1.43, H3-K9-HMTase 4, Histone H3-K9 methyltransferase 4, histone-lysine N-methyltransferase SETDB1, histone-lysine N-methyltransferase, H3lysine-9 specific 4, KG1T, KIAA0067H3-K9-HMTase4, KMT1EERG-associated protein with a SET domain, ESET, Lysine N-methyltransferase 1E, SET domain bifurcated 1, SET domain, bifurcated 1 | |
| The immunogen recognized by this antibody maps to a region between residues 1125 and 1175 of human SET Domain, Bifurcated 1 using the numbering given in entry NP_036564.2 (GeneID 9869). | |
| 0.1 mL | |
| Epigenetics, Transcription Factors and Regulators | |
| 9869 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction