missing translation for 'onlineSavingsMsg'
Learn More

SETDB2 Antibody [Alexa Fluor« 405], Novus Biologicals Biologicals™

Product Code. 30512528 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30512528 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30512528 Supplier Novus Biologicals Supplier No. NBP338422AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SETDB2 Polyclonal antibody specifically detects SETDB2 in Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen SETDB2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias C13orf4, chromosome 13 open reading frame 4, chronic lymphocytic leukemia deletion region 8, Chronic lymphocytic leukemia deletion region gene 8 protein, CLLD8DKFZp761J1217, DKFZp586I0123, EC 2.1.1.43, histone-lysine N-methyltransferase SETDB2, KMT1FCLLL8, Lysine N-methyltransferase 1F, SET domain bifurcated 2, SET domain, bifurcated 2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 148-378 of human SETDB2 (NP_114121.2).,, Sequence:, SGACLMKMPLNLKGENPLQLPIKCHFQRRHAKTNSHSSALHVSYKTPCGRSLRNVEEVFRYLLETECNFLFTDNFSFNTYVQLARNYPKQKEVVSDVDISNGVESVPISFCNEIDSRKLPQFKYRKTVWPRAYNLTNFSSMFTDSCDCSEGCIDITKCACLQLTARNAKTSPLSSDKITTGYKYKRLQRQIPTGIYECSLLCKCNRQLCQNRVVQHGPQVRLQVFKTEQKG
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83852
Target Species Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.