Learn More
Novus Biologicals™ SEZ6/BSRP-C Antibody (Abbv-011) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28127-100ug
 View more versions of this product
Product Code. 30044418
  • € 349.00 / 100µg
Description
SEZ6/BSRP-C Monoclonal antibody specifically detects SEZ6/BSRP-C in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SEZ6/BSRP-C
Monoclonal
Unconjugated
Q53EL9
Human
Protein A purified
RUO
124925
Human
Purified
Flow Cytometry, ELISA, Functional Assay
Abbv-011
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
hSEZ-6, seizure related 6 homolog (mouse)
SEZ6
100 μg
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.