missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEZ6L2/BSRP-A Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97264AF532
This item is not returnable.
View return policy
Description
SEZ6L2/BSRP-A Polyclonal antibody specifically detects SEZ6L2/BSRP-A in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SEZ6L2/BSRP-A | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 26470 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Alexa Fluor 532 | |
| seizure 6-like protein 2, seizure related 6 homolog (mouse)-like 2, UNQ1903/PRO4349 | |
| Produced in rabbits immunized with E. coli-derived Human SEZ6L2/BSRP-A fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction