Learn More
SF-1/NR5A1/Steroidogenic Factor 1 Antibody (SF1/8000R) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-20854
Product Code. 18614254
  • € 592.00 / 100µg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
SF-1/NR5A1/Steroidogenic Factor 1 Monoclonal antibody specifically detects SF-1/NR5A1/Steroidogenic Factor 1 in Human, Rat samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spécification
Spécification
SF-1/NR5A1/Steroidogenic Factor 1
Monoclonal
1 mg/mL
Immunohistochemistry-Paraffin 1-2 μg/mL
AD4BPSTF-1, Adrenal 4-binding protein, ELP, FTZ1adrenal 4 binding protein, FTZF1nuclear receptor AdBP4, Fushi tarazu factor homolog 1, Nuclear receptor subfamily 5 group A member 1, nuclear receptor subfamily 5, group A, member 1, SF-1POF7, SF1steroidogenic factor 1, Steroid hormone receptor Ad4BP, steroidogenic factor-1
Recombinant fragment (around aa200-400) of human SF-1/NR5A1/Steroidogenic Factor 1 protein (exact sequence is proprietary)
100 μg
Cell Biology
2516
Store at -20°C to -70°C. Avoid freeze/thaw cycles.
IgG κ
Immunohistochemistry (Paraffin)
SF1/8000R
Unconjugated
10 mM PBS
Rabbit
Protein A or G purified
RUO
Primary
Human, Rat