missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SF-1/NR5A1/Steroidogenic Factor 1 Antibody (SF1/8000R), DyLight 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20854V
This item is not returnable.
View return policy
Description
SF-1/NR5A1/Steroidogenic Factor 1 Monoclonal antibody specifically detects SF-1/NR5A1/Steroidogenic Factor 1 in Human, Rat samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SF-1/NR5A1/Steroidogenic Factor 1 | |
| Monoclonal | |
| DyLight 405 | |
| AD4BPSTF-1, Adrenal 4-binding protein, ELP, FTZ1adrenal 4 binding protein, FTZF1nuclear receptor AdBP4, Fushi tarazu factor homolog 1, Nuclear receptor subfamily 5 group A member 1, nuclear receptor subfamily 5, group A, member 1, SF-1POF7, SF1steroidogenic factor 1, Steroid hormone receptor Ad4BP, steroidogenic factor-1 | |
| Recombinant fragment (around aa200-400) of human SF-1/NR5A1/Steroidogenic Factor 1 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cell Biology | |
| 2516 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| SF1/8000R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction