missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SF3A1 Monoclonal Antibody (OTI1B5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800496S
This item is not returnable.
View return policy
Description
SF3A1 Monoclonal antibody specifically detects SF3A1 in Human samples. It is validated for Western BlotSpecifications
| SF3A1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P04271 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6285 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1B5 | |
| Unconjugated | |
| S100B | |
| 1200014H24Rik; 5930416L09Rik; AI159724; pre-mRNA processing 21; pre-mRNA splicing factor SF3a (120 kDa subunit); PRP21; PRPF21; SAP 114; SAP114; Sf3a1; sf3a1 protein; SF3A120; si:dz150f13.2; spliceosome-associated protein 114; splicing factor 3 subunit 1; splicing factor 3A subunit 1; splicing factor 3a, subunit 1; splicing factor 3a, subunit 1, 120kD; splicing factor 3a, subunit 1, 120kDa; Unknown (protein for MGC:65786); wu:fc38e01; wu:fc50a11; wu:fj37c05; zgc:65786 | |
| Full length human recombinant protein of human S100B produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction