missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SF3A3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29283
This item is not returnable.
View return policy
Description
SF3A3 Polyclonal antibody specifically detects SF3A3 in Human,Mouse samples. It is validated for Immunohistochemistry
Specifications
| SF3A3 | |
| Polyclonal | |
| Tris-buffered Saline 0.1% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10946 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunohistochemistry | |
| Unconjugated | |
| PRP9, PRPF9, SAP 61, SAP61splicing factor 3a, subunit 3, 60kD, SF3a60pre-mRNA splicing factor SF3a (60kD), spliceosome associated protein 61, Spliceosome-associated protein 61, splicing factor 3A subunit 3, splicing factor 3a, subunit 3, 60kDa | |
| Between 451 and 501 | |
| 25 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction