missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SF3A3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556909
This item is not returnable.
View return policy
Description
Immunogen sequence: LGWDGKPIPY WLYKLHGLNI NYNCEICGNY TYRGPKAFQR HFAEWRHAHG MRCLGIPNTA HFANVTQIED AVSLWAKLKL QK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
This gene encodes subunit 3 of the splicing factor 3a protein complex. The splicing factor 3a heterotrimer includes subunits 1, 2 and 3 and is necessary for the in vitro conversion of 15S U2 snRNP into an active 17S particle that performs pre-mRNA splicing. Subunit 3 interacts with subunit 1 through its amino-terminus while the zinc finger domain of subunit 3 plays a role in its binding to the 15S U2 snRNP. This gene has a pseudogene on chromosome 20.
Specifications
| SF3A3 | |
| Polyclonal | |
| Unconjugated | |
| SF3A3 | |
| 4930512K19Rik; 60kDa; hypothetical protein LOC523250; LRRGT00093; pre-mRNA splicing factor SF3a (60kD); PRP9; PRPF9; SAP 61; Sap61; SF3A3; SF3a60; spliceosome associated protein 61; spliceosome-associated protein 61; splicesome-associated factor 61; splicing factor 3A subunit 3; splicing factor 3a subunit 3-like protein; splicing factor 3a, subunit 3; splicing factor 3a, subunit 3, 60kD; splicing factor 3a, subunit 3, 60kDa; wu:fi15a06; wu:fj94f01; zgc:110227 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10946 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q12874 | |
| SF3A3 | |
| Recombinant protein corresponding to Human SF3A3. Recombinant protein control fragment (Product #RP-102613). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction