missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SF3B1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541724
This item is not returnable.
View return policy
Description
Peptide sequence: MAKIAKTHED IEAQIREIQG KKAALDEAQG VGLDSTGYYD QEIYGGSDSR Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
This gene encodes subunit 1 of the splicing factor 3b protein complex. Splicing factor 3b, together with splicing factor 3a and a 12S RNA unit, forms the U2 small nuclear ribonucleoproteins complex. The splicing factor 3b/3a complex binds pre-mRNA upstream of the intron's branch site in a sequence independent manner and may anchor the U2 snRNP to the pre-mRNA. Splicing factor 3b is also a component of the minor U12-type spliceosome. The carboxy-terminal two-thirds of subunit 1 have 22 non-identical, tandem HEAT repeats that form rod-like, helical structures. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| SF3B1 | |
| Polyclonal | |
| Unconjugated | |
| SF3B1 | |
| 155kDa; 2810001M05Rik; AA409119; Hsh155; LOW QUALITY PROTEIN: splicing factor 3B subunit 1; MDS; pre-mRNA processing 10; pre-mRNA splicing factor SF3b, 155 kDa subunit; pre-mRNA-splicing factor SF3b 155 kDa subunit; PRP10; PRPF10; SAP 155; SAP155; SF3B1; SF3b155; spliceosome-associated protein 155; Splicing factor 3B subunit 1; splicing factor 3b subunit 1, 155kDa; splicing factor 3b, subunit 1; splicing factor 3b, subunit 1, 155 kDa; splicing factor 3b, subunit 1, 155kD; splicing factor 3b, subunit 1, 155kDa; TA-8; Targ4; transforming growth factor alpha regulated gene 4; tst; wu:fb99f09 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 23451 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O75533 | |
| SF3B1 | |
| Synthetic peptide directed towards the N-terminal of human SF3B1 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction