missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SF3B4 Monoclonal Antibody (3A1)

Mouse Monoclonal Antibody
€ 491.00 - € 1704.00
Specifications
| Antigen | SF3B4 |
|---|---|
| Clone | 3A1 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17875107
|
Invitrogen™
MA547309 |
500 μL |
€ 1704.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17815227
![]() |
Invitrogen™
MA547407 |
100 μL |
€ 491.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SF3B4 Monoclonal Antibody for Western Blot, ICC/IF, IHC
This gene encodes one of four subunits of the splicing factor 3B. The protein encoded by this gene cross-links to a region in the pre-mRNA immediately upstream of the branchpoint sequence in pre-mRNA in the prespliceosomal complex A. It also may be involved in the assembly of the B, C and E spliceosomal complexes. In addition to RNA-binding activity, this protein interacts directly and highly specifically with subunit 2 of the splicing factor 3B. This protein contains two N-terminal RNA-recognition motifs (RRMs), consistent with the observation that it binds directly to pre-mRNA.Specifications
| SF3B4 | |
| 1 mg/mL | |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 5mM sodium azide | |
| Q15427, Q6AYL5, Q8QZY9 | |
| 10262, 107701, 295270 | |
| Full length recombinant human SF3B4 expressed in and purified from E. coli. | |
| Antibody |
| 3A1 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG2b | |
| Human, Mouse, Rat | |
| Sf3b4 | |
| 49kD; AFD1; chunp6902; fb36a04; Hsh49; MGC10828; pre-mRNA-splicing factor SF3b 49 kDa subunit; SAP 49; Sap49; SAP49MGC10828; sf3b4; SF3b49; SF3b50; spliceosomal protein; spliceosome-associated protein (U2 snRNP); spliceosome-associated protein 49; Splicing factor 3B subunit 4; splicing factor 3b, subunit 4; splicing factor 3b, subunit 4, 49kD; splicing factor 3b, subunit 4, 49kDa; Unknown (protein for IMAGE:8120437); wu:fb36a04 | |
| Sf3b4 | |
| Primary | |
| Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title