Learn More
SF3B5 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-18419AF532
Product Code. 18508307
  • € 658.00 / 0.10mL
Description
SF3B5 Polyclonal antibody specifically detects SF3B5 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SF3B5
Polyclonal
Western Blot
MGC3133, Pre-mRNA-splicing factor SF3b 10 kDa subunit, SF3b10, SF3b5, splicing factor 3B subunit 5, splicing factor 3b, subunit 5, 10kDa, Ysf3
Recognizes region between residues Between Between 1 and 50..
0.1 mL
Primary
Human, Mouse
Purified
Western Blot
Alexa Fluor 532
50mM Sodium Borate
Rabbit
Affinity purified
RUO
83443
Store at 4°C in the dark.