Learn More
Motic™ Photo Adapter 2Xslr
Suitable for SMZ-168, SMZ 143-N2GG microscopes. Motic™ Photo Adapter 2Xslr allows you to connect a 35mm camera.
Brand: Motic™ 11143181
Additional Details : Weight : 0.01000kg
Description
- Suitable for SMZ-168, SMZ 143-N2GG microscopes.
- Connects to 35mm camera.
- A bayonet adapter is also required.
Specifications
Photo Adapter | |
SMZ-168, SMZ 143-N2GG microscopes | |
2X |