missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SFMBT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30523
This item is not returnable.
View return policy
Description
SFMBT1 Polyclonal antibody specifically detects SFMBT1 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| SFMBT1 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 51460 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| DKFZp434L243, Renal ubiquitous protein 1, RU1scm-like with four MBT domains protein 1, Scm-like with four mbt domains 1, Scm-related gene containing four mbt domains, Scm-related gene product containing four mbt domains, SFMBT | |
| between 816 and 866 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction