missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFMBT1 Antibody (RAB-C396), Alexa Fluor™ 700 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20147AF700
This item is not returnable.
View return policy
Description
SFMBT1 Monoclonal antibody specifically detects SFMBT1 in Human samples. It is validated for ELISA
Specifications
| SFMBT1 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| DKFZp434L243, Renal ubiquitous protein 1, RU1scm-like with four MBT domains protein 1, Scm-like with four mbt domains 1, Scm-related gene containing four mbt domains, Scm-related gene product containing four mbt domains, SFMBT | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing SFMBT1 protein under non-denaturing conditions. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| RAB-C396 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 51460 | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction