missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SFMBT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583273
This item is not returnable.
View return policy
Description
Immunogen sequence: DFWCDVVIAD LHPVGWCTQN NKVLMPPDAI KEKYTDWTEF LIRDLTGSRT APANLLEGPL RGKGPIDLIT VGSLIELQDS QN Highest antigen sequence indentity to the following orthologs: Rat - 70%, Mouse - 91%.
The SFMBT2 (Scm-like with four MBT domains 2) gene shares high similarity with the Drosophila Scm (sex comb on midleg) gene. Sfmbt2 is a Polycomb group (PcG) gene that maps to the proximal region of Chromosome 2, and is a putative imprinted gene. Sfmbt2 is the first imprinted gene within this region to be identified. Studies indicate that six different translocations involving proximal chromosome 2 results in lethality when present as a maternal uniparental duplication.
Specifications
| SFMBT2 | |
| Polyclonal | |
| Unconjugated | |
| SFMBT2 | |
| D2Wsu23e; D330030P06Rik; KIAA1617; Scm-like with 4 MBT domains protein 2; Scm-like with four mbt domains 2; scm-like with four MBT domains protein 2; Scm-related gene containing four mbt domain 2; Scm-related gene containing four mbt domains 2; SFMBT2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57713 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5VUG0 | |
| SFMBT2 | |
| Recombinant protein corresponding to Human SFMBT2. Recombinant protein control fragment (Product #RP-97214). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction