missing translation for 'onlineSavingsMsg'
Learn More
Learn More
sFRP-1 Antibody (024), DyLight 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89596G
This item is not returnable.
View return policy
Description
sFRP-1 Monoclonal antibody specifically detects sFRP-1 in Human samples. It is validated for ELISA
Specifications
| sFRP-1 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 024 | |
| ELISA | |
| FRP1, FRP-1FrzA, FRPSARP-2, SARP2sFRP-1, Secreted apoptosis-related protein 2, secreted frizzled-related protein 1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human sFRP-1 (NP_003003.3; Met1-Lys314). | |
| 0.1 mL | |
| Apoptosis | |
| 6422 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction