missing translation for 'onlineSavingsMsg'
Learn More
Learn More
sFRP-2 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18244JF646
This item is not returnable.
View return policy
Description
sFRP-2 Polyclonal antibody specifically detects sFRP-2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| sFRP-2 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry | |
| FRP2, FRP-2secreted apoptosis related protein 1, SARP-1, SARP1sFRP-2, SDF-5, Secreted apoptosis-related protein 1, secreted frizzled-related protein 2 | |
| Synthetic peptide from the C-terminal of Human Secreted frizzled-related protein 2 (aa. 195-295) | |
| 0.1 mL | |
| Stem Cell Markers | |
| 6423 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction