missing translation for 'onlineSavingsMsg'
Learn More
Learn More
sFRP-2 Antibody (OTI6C2), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74151F
This item is not returnable.
View return policy
Description
sFRP-2 Monoclonal specifically detects sFRP-2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
| sFRP-2 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| SFRP2 | |
| Recombinant protein fragment corresponding to amino acids 25-295 of human sFRP-2 (NP_003004) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Immunohistochemistry | |
| OTI6C2 | |
| Immunohistochemistry | |
| FRP2, FRP-2secreted apoptosis related protein 1, SARP-1, SARP1sFRP-2, SDF-5, Secreted apoptosis-related protein 1, secreted frizzled-related protein 2 | |
| Mouse | |
| Affinity Purified | |
| Stem Cell Markers | |
| 6423.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction