Learn More
sFRP-5 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94775-0.1ml
This item is not returnable.
View return policy
Description
sFRP-5 Polyclonal antibody specifically detects sFRP-5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| sFRP-5 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/ Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin | |
| frizzled-related protein 1b, FRP-1b, FRP1B, SARP3, SARP-3, secreted apoptosis related protein 3, secreted apoptosis-related protein 3, secreted frizzled-related protein 5, sFRP-5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 30-317 of human SFRP5 (NP_003006.2). EEYDYYGWQAEPLHGRSYSKPPQCLDIPADLPLCHTVGYKRMRLPNLLEHESLAEVKQQASSWLPLLAKRCHSDTQVFLCSLFAPVCLDRPIYPCRSLCEAVRAGCAPLMEAYGFPWPEMLHCHKFPLDNDLCIAVQFGHLPATAPPVTKICAQCEMEHSADGLMEQMCSSDFVVKMRIKEIKIENGDRKLIGAQKKKKLLKPGPLKRKDTKRLVLHMKNGAGCPCPQLDSLAGSFLVMGRKVDGQLLLMAVYRWDKKNKEMKFAVKFMFSYPCSLYYPFFYGAAEPH | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Stem Cells | |
| 6425 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |