Learn More
Invitrogen™ SFRS12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589104
Description
Immunogen sequence: MTSLMPGAGL LPIPTPNPLT TLGVSLSSLG AIPAAALDPN IATLGEIPQP PLMGNVDPSK IDEIRRTVYV GNLNSQTTTA DQLLEFFKQV GEVKFVRMAG DETQPTRFAF VEFADQNSVP RALAFNGVMF GDRPLKINHS NNAIVKPPEM TPQAAAKELE EVMKRVREAQ SFISAAIEPE; Positive Samples: MCF-7, HeLa; Cellular Location: Nucleus.
SFRS12 belongs to the superfamily of serine/arginine-rich splicing factors. It modulates splice site selection by regulating the activities of other SR proteins.
Specifications
| SFRS12 | |
| Polyclonal | |
| Unconjugated | |
| SREK1 | |
| 8430401B01; AI450757; AI462342; serine/arginine-rich splicing factor 12; serine/arginine-rich-splicing regulatory protein 86; serine-arginine-rich splicing regulatory protein 508; serine-arginine-rich splicing regulatory protein 86; serine-arginine-rich-splicing regulatory protein 86; SFRS12; Splicing factor, arginine/serine-rich 12; splicing regulatory glutamic acid and lysine rich protein 1; splicing regulatory glutamic acid/lysine-rich protein 1; splicing regulatory glutamine/lysine-rich protein 1; Splicing regulatory protein 508; Srek1; SR-related protein of 86 kDa; SRrp508; Srrp86; Srsf12 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 140890, 218543 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.48 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8BZX4, Q8WXA9 | |
| SREK1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human SREK1 (NP_6319071). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.