missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SFRS19 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561345
This item is not returnable.
View return policy
Description
Immunogen sequence: SRKVKLQSKV AVLIREGVSS TTPAKDAASA GLGSIGVKFS RDRESRSPFL KPDERAPTEM AKAAPGSTKP KKTKVKAKAG AKKTKGTKGK TKPSKT Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
Splicing factor that seems to antagonize SR proteins in pre-mRNA splicing regulation. Also binds the repetitive C-terminal domain (CTD) of RNA polymerase II; may act with other CTD binding proteins to link transcription and pre-mRNA processing.
Specifications
| SFRS19 | |
| Polyclonal | |
| Unconjugated | |
| SCAF1 | |
| AI480556; CTD-binding SR-like protein rA1; CTD-binding SR-like rA1; SCAF; Scaf1; Serine arginine-rich pre-mRNA splicing factor SR-A1; SFRS19; Splicing factor, arginine/serine-rich 19; SRA1; Sr-a1; SR-related and CTD-associated factor 1; SR-related CTD associated factor 1; SR-related CTD-associated factor 1; SR-related C-terminal domain-associated factor 1; SR-related-CTD-associated factor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 58506 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H7N4 | |
| SCAF1 | |
| Recombinant protein corresponding to Human SFRS19. Recombinant protein control fragment (Product #RP-99013). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction