missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA Polymerase epsilon catalytic subunit A, Mouse anti-Human,Mouse,Mustelid,Primate, DyLight 350, Clone: 3C5.1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-115UV-0.1ML
This item is not returnable.
View return policy
Description
DNA Polymerase epsilon catalytic subunit A Monoclonal specifically detects DNA Polymerase epsilon catalytic subunit A in Human, Mouse, Mustelid, Primate, Drosophila (Negative) samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| DNA Polymerase epsilon catalytic subunit A | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| POLE | |
| Human DNA Polymerase epsilon purified from HeLa cells. | |
| 0.1 mL | |
| Chromatin Research, DNA Polymerases, DNA Repair | |
| 5426 | |
| Human, Mouse, Mustelid, Primate, Drosophila (Negative) | |
| Purified |
| Western Blot, Immunofluorescence | |
| 3C5.1 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| DKFZp434F222, DNA polymerase epsilon catalytic subunit A, DNA polymerase II subunit A, EC 2.7.7, EC 2.7.7.7, POLE1FLJ21434, polymerase (DNA directed), epsilon | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This is specific to the human DNA pol epsilon and does not recognize DNA pol alpha, beta or delta. | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction