missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SFRS8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA537068
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 105 kDa. Purity is >95% by SDS-PAGE.
Signal-induced proliferation associated-like protein 2 (SIPA1L2) is a member of the SIPA1 family of RapGAPs. Little is known of the role of the SIPA1L2 protein, but recent studies of SIPA indicate that its deregulation can cause myeloproliferative stem cell disorders in mice and increased metastases in human cancers. Other studies suggest SIPA1L1 may play important roles in embryo development and control of cell proliferation. Based on the amount of homology between SIPA family members, it is likely that SIPA1L2 plays a role in embryo development and cell proliferation, possibly including oncogenesis.
Specifications
| SFRS8 | |
| Polyclonal | |
| Unconjugated | |
| SFSWAP | |
| 1190005N23Rik; 6330437E22Rik; AI197402; AW212079; serine/arginine-rich splicing factor 8; Sfrs8; SFSWAP; splicing factor SWAP homolog; splicing factor, arginine/serine-rich 8; splicing factor, arginine/serine-rich 8 (suppressor-of-white-apricot homolog, Drosophila); splicing factor, arginine/serine-rich 8 (suppressor-of-white-apricot, Drosophila homolog); splicing factor, suppressor of white-apricot family; splicing factor, suppressor of white-apricot homolog; splicing factor, suppressor of white-apricot homolog (Drosophila); Srsf8; Suppressor of white apricot protein homolog; SWAP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 231769, 304431, 6433 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| D3ZTQ1, Q12872, Q3USH5 | |
| SFSWAP | |
| Synthetic peptide corresponding to amino acids 251-296 of human SFRS8. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction