missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFT2B Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98132DL594
This item is not returnable.
View return policy
Description
SFT2B Polyclonal antibody specifically detects SFT2B in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SFT2B | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 375035 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| dJ747L4.C1.2, FLJ34085, SFT2 domain containing 2, SFT2 domain-containing protein 2, UNQ512, vesicle transport protein SFT2B | |
| Produced in rabbits immunized with E. coli-derived Human SFT2B fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction