missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SFTPC Monoclonal Antibody (5E6A9)
GREENER_CHOICE

Product Code. 15358130
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15358130 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15358130 Supplier Invitrogen™ Supplier No. MA517172

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA5-17172 targets SFTPC in indirect ELISA and WB applications and shows reactivity with Human samples. The MA5-17172 immunogen is purified recombinant fragment of human SFTPC (amino acids: 60-180 ) expressed in E. Coli. MA5-17172 detects SFTPC which has a predicted molecular weight of approximately 21kDa.

This gene encodes the pulmonary-associated surfactant protein C (SPC), an extremely hydrophobic surfactant protein essential for lung function and homeostasis after birth. Pulmonary surfactant is a surface-active lipoprotein complex composed of 90% lipids and 10% proteins which include plasma proteins and apolipoproteins SPA, SPB, SPC and SPD. The surfactant is secreted by the alveolar cells of the lung and maintains the stability of pulmonary tissue by reducing the surface tension of fluids that coat the lung. Multiple mutations in this gene have been identified, which cause pulmonary surfactant metabolism dysfunction type 2, also called pulmonary alveolar proteinosis due to surfactant protein C deficiency, and are associated with interstitial lung disease in older infants, children, and adults. Alternatively spliced transcript variants encoding different protein isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFTPC
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5E6A9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Sftpc
Gene Accession No. P11686
Gene Alias BRICD6; BRICHOS domain containing 6; LOW QUALITY PROTEIN: pulmonary surfactant-associated protein C; pro-SpC; PSP C; PSP-C; pulmonary surfactant apoprotein-2 SP-C; pulmonary surfactant associated protein C; pulmonary surfactant protein C; pulmonary surfactant-associated protein C; Pulmonary surfactant-associated proteolipid SPL(Val); SFTP 2; SFTP2; Sftp-2; Sftpc; SMDP2; SP C; SP5; SPC; SP-C; surfactant associated protein C; surfactant protein C; Surfactant pulmonary-associated protein C; Surfactant, pulmonary-associated protein C; surfactant-associated protein; type I SP-C; type I surfactant protein C; type II SP-C; type II surfactant protein C
Gene Symbols Sftpc
Host Species Mouse
Immunogen Purified recombinant fragment of human SFTPC (amino acids: 60-180 ) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6440
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.