missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFTS Virus HB29 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41155AF532
This item is not returnable.
View return policy
Description
SFTS Virus HB29 Polyclonal antibody specifically detects SFTS Virus HB29 in Virus samples. It is validated for ELISA
Specifications
| SFTS Virus HB29 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
| ELISA | |
| Alexa Fluor 532 | |
| B303_sMgp1, Membrane glycoprotein polyprotein | |
| Antibody was raised against an 18 amino acid synthetic peptide near the center of human SFTS Virus HB29 Membrane Glycoprotein. The immunogen is located within amino acids 570 - 620 of SFTS Virus HB29. Amino Acid Squence: ITTGRALLPAVNPGQEA | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| 13231111 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction