missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFTS Virus HB29 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41156H
This item is not returnable.
View return policy
Description
SFTS Virus HB29 Polyclonal antibody specifically detects SFTS Virus HB29 in Virus samples. It is validated for Western Blot, ELISA
Specifications
| SFTS Virus HB29 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
| Western Blot, ELISA | |
| HRP | |
| B303_sMgp1, Membrane glycoprotein polyprotein | |
| Antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human SFTS Virus HB29 Membrane Glycoprotein. The immunogen is located within the last 50 amino acids of SFTS Virus HB29. Amino Acid Squence: KLGTKQVFRSRTKLA | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| 13231111 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction