missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROR2 Antibody (231512), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2064B
This item is not returnable.
View return policy
Description
ROR2 Monoclonal antibody specifically detects ROR2 in Human samples. It is validated for Western Blot
Specifications
| ROR2 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 231512 | |
| Western Blot | |
| BDB, BDB1, EC 2.7.10.1, MGC163394, neurotrophic tyrosine kinase receptor-related 2, Neurotrophic tyrosine kinase, receptor-related 2, NTRKR2tyrosine-protein kinase transmembrane receptor ROR2, receptor tyrosine kinase-like orphan receptor 2 | |
| Mouse myeloma cell line NS0-derived recombinant human ROR2, Val28-Gly403, Accession # AAA60276 | |
| 0.1 mL | |
| Protein Kinase, Wnt Signaling Pathway | |
| 4920 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction